SY3H3249F9BF Synthonix Corporation

Methyl N-[2-[3,5-bis[trifluoromethyl]phenyl]-1-methyl-2-oxo-ethyl]carbamate

  • MDL number MFCD19707643
  • Cas Number : 1363382-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7762.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7762.html
hstatements H302 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart