SY3H3249F9C1 Synthonix Corporation

Methyl-4-oxotetrahydro-2H-thiopyran-3-carboxylate

  • MDL number MFCD00203485
  • Cas Number : 4160-61-6
  • Molecular Weight : 174.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8265.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/8265.html
pictograms GHS05, GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart