SY3H3249F9C2 Synthonix Corporation

Methyl 2-(Boc-amino)-2-(oxetan-3-yl)acetate

  • MDL number MFCD22628746
  • Cas Number : 1416323-08-4
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8437.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/8437.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart