SY3H3249F9CB Synthonix Corporation

1-(((4-Nitrophenoxy)carbonyl)oxy)ethyl isobutyrate

  • MDL number MFCD22571566
  • Cas Number : 194995-47-6
  • Molecular Weight : 297.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/32162.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/32162.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart