SY3H3249F9DF Synthonix Corporation

1-Oxa-5-azaspiro[2.5]octane-5-carboxylic acid tert-butyl ester

  • MDL number MFCD07779386
  • Cas Number : 276872-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/1986.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1986.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart