SY3H3249F9E1 Synthonix Corporation

1-Methylazepan-4-amine

  • MDL number MFCD17015865
  • Cas Number : 933741-93-6
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0131.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/0131.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart