SY3H3249F9E2 Synthonix Corporation

2-Oxa-6-azaspiro[3.3]heptane hemioxalate

  • MDL number MFCD16652332
  • Cas Number : 1045709-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/5816.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5816.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart