SY3H3249F9E5 Synthonix Corporation

1-Oxa-6-azaspiro[3.4]octane

  • MDL number MFCD18325163
  • Cas Number : 130906-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/5948.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5948.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart