SY3H3249F9EC Synthonix Corporation

1-Oxa-6-azaspiro[3.5]nonane hemioxalate

  • MDL number MFCD27923648
  • Cas Number : 1523606-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8371.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8371.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart