SY3H3249F9F2 Synthonix Corporation

3-Pyrrolidineacetic acid, 1-[(1,1-dimethylethoxy)carbonyl]-, methyl ester

  • MDL number MFCD18910910
  • Cas Number : 890849-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/0250.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0250.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart