SY3H3249F9F6 Synthonix Corporation

1-tert-butyl 3-Ethyl 4-oxopyrrolidine-1,3-dicarboxylate

  • MDL number MFCD06656201
  • Cas Number : 146256-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0562.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0562.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart