SY3H3249F9FA Synthonix Corporation

2-(3-Piperidyl)acetonitrile hydrochloride

  • MDL number MFCD19443966
  • Cas Number : 5562-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/0995.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0995.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart