SY3H3249F9FF Synthonix Corporation

1-Cbz-piperidine-3-acetic acid

  • MDL number No MDL found
  • Cas Number : 86827-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10870.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10870.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart