SY3H3249FA05 Synthonix Corporation

Pyrrolidin-3-ylmethanol hydrochloride

  • MDL number MFCD09800469
  • Cas Number : 644971-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1155.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1155.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart