SY3H3249FA07 Synthonix Corporation

4-PHENYL-2,3-DIHYDRO-1,3-THIAZOL-2-ONE

  • MDL number No MDL found
  • Cas Number : 63615-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11961.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11961.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart