SY3H3249FA0F Synthonix Corporation

2-Piperazin-1-yl-propan-1-ol

  • MDL number No MDL found
  • Cas Number : 71850-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13371.html

Safety Information

MSDS https://synthonix.com/13371.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart