SY3H3249FA64 Synthonix Corporation

2-(Piperazin-1-yl)-4-(trifluoromethyl)pyrimidine

  • MDL number No MDL found
  • Cas Number : 179756-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/38202.html

Safety Information

MSDS https://synthonix.com/38202.html
Availability details for
Quantity
  • Availability
  • Add to Cart