SY3H3249FA8F Synthonix Corporation

1-Pyrrolidinecarboxylic acid, 3-ethynyl-,1,1-dimethyl ester

  • MDL number MFCD10698160
  • Cas Number : 287193-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7677.html

Safety Information

MSDS https://synthonix.com/7677.html
hstatements H302 ,
Availability details for
Quantity
  • Availability
  • Add to Cart