SY3H3249FAA3 Synthonix Corporation

1-Methyl-2-oxo-piperidine-4-carboxylic acid

  • MDL number MFCD09863345
  • Cas Number : 1000932-09-1
  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0207.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/0207.html
pictograms GHS07
hstatements H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart