SY3H3249FAA5 Synthonix Corporation

Tetrahydro-3-furanylmethyl bromide

  • MDL number MFCD08234790
  • Cas Number : 165253-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0399.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0399.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart