SY3H3249FAAF Synthonix Corporation

Tetrahydro-2H-thiopyran-3-carboxylic acid

  • MDL number MFCD14584577
  • Cas Number : 161404-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1163.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1163.html
hstatements H302 , H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart