SY3H3249FAB1 Synthonix Corporation

4-Boc-2-(4-(trifluoromethyl)phenyl)piperazine

  • MDL number No MDL found
  • Cas Number : 886767-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11783.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11783.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart