SY3H3249FAB5 Synthonix Corporation

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-ene-1-carbonitrile

  • MDL number No MDL found
  • Cas Number : 1310384-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/12110.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/12110.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart