SY3H3249FAB6 Synthonix Corporation

Tetrahydrofuran-3-carbonitrile

  • MDL number No MDL found
  • Cas Number : 14631-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/12452.html

Safety Information

MSDS https://synthonix.com/12452.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart