SY3H3249FABA Synthonix Corporation

1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-boronic acid pinacol ester

  • MDL number No MDL found
  • Cas Number : 956388-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14464.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/14464.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart