SY3H3249FABF Synthonix Corporation

Tetrahydro-2H-pyran-3-carboxylic acid hydrazide

  • MDL number No MDL found
  • Cas Number : 59293-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/15052.html

Safety Information

MSDS https://synthonix.com/15052.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart