SY3H3249FADB Synthonix Corporation

1,1,1-trifluoro-2-(piperidin-4-yl)propan-2-ol

  • MDL number No MDL found
  • Cas Number : 1227068-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/29566.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/29566.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart