SY3H3249FAE5 Synthonix Corporation

1-Methyl-3-(aminomethyl)piperidine

  • MDL number MFCD06738902
  • Cas Number : 14613-37-7
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0254.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/0254.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart