SY3H3249FB20 Synthonix Corporation

1-(Tert-butoxycarbonyl)-4,4-difluoropiperidine-3-carboxylic acid

  • MDL number MFCD18909829
  • Cas Number : 1303974-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/7119.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7119.html
hstatements H302 , H315 , H317 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart