SY3H3249FB33 Synthonix Corporation

5-Cbz-1-oxa-5-azaspiro[2.4]heptane

  • MDL number No MDL found
  • Cas Number : 790704-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11563.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11563.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart