SY3H3249FB3B Synthonix Corporation

2-Oxo-pyrrolidine-3-carboxylic acid ethyl ester

  • MDL number No MDL found
  • Cas Number : 36821-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13453.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/13453.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart