SY3H3249FB40 Synthonix Corporation

2-Boc-octahydropyrazino[1,2-a]pyrazine

  • MDL number No MDL found
  • Cas Number : 1159825-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13687.html

Safety Information

MSDS https://synthonix.com/13687.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart