SY3H3249FB41 Synthonix Corporation

tert-butyl 3-Oxo-1-phenyl-2,7-diazaspiro[3.5]nonane-7-carboxylate

  • MDL number MFCD18089860
  • Cas Number : 1013033-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/1369.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1369.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart