SY3H3249FB44 Synthonix Corporation

6-Oxa-1-aza-spiro[3.5]nonane

  • MDL number No MDL found
  • Cas Number : 1214875-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14350.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/14350.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart