SY3H3249FB46 Synthonix Corporation

9-Boc-6-oxa-1,9-diazaspiro[3.6]decane

  • MDL number No MDL found
  • Cas Number : 1250998-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14375.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/14375.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart