SY3H3249FB49 Synthonix Corporation

Methyl 6-oxopiperidine-3-carboxylate

  • MDL number MFCD17016096
  • Cas Number : 958991-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0586.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0586.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart