SY3H3249FB54 Synthonix Corporation

2-Oxopiperidine-4-carboxylic acid

  • MDL number MFCD13659408
  • Cas Number : 24537-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/1724.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1724.html
hstatements H302 , H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart