SY3H3249FB66 Synthonix Corporation

2-(3-Oxocyclopentyl)isoindoline-1,3-dione

  • MDL number MFCD18910252
  • Cas Number : 1029691-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37318.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37318.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart