SY3H3249FB68 Synthonix Corporation

tert-Butyl 3-(2-(tert-butoxy)-2-oxoethyl)-4-oxopiperidine-1-carboxylate

  • MDL number MFCD11111810
  • Cas Number : 1010814-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37702.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37702.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart