SY3H3249FB69 Synthonix Corporation

N-(2-Oxoethyl)phthalimide

  • MDL number MFCD00023080
  • Cas Number : 2913-97-5
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)acetaldehyde
Inchi Code 1S/C10H7NO3/c12-6-5-11-9(13)7-3-1-2-4-8(7)10(11)14/h1-4,6H,5H2
Inchi Key LMRDBJZQDUVCQH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/37866.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37866.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart