SY3H3249FB7A Synthonix Corporation

1-Oxa-4,8-diazaspiro[5.5]undecane-8-carboxylic acid tert-butyl ester

  • MDL number MFCD11227062
  • Cas Number : 1160247-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6033.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6033.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart