SY3H3249FB7E Synthonix Corporation

3-Oxa-8-azaspiro[3.5]nonane-8-carboxylic acid tert-butyl ester

  • MDL number MFCD18642709
  • Cas Number : 1272412-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/6166.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6166.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart