SY3H3249FB7F Synthonix Corporation

2-Oxetanemethanamine

  • MDL number MFCD17214171
  • Cas Number : 882402-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6187.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6187.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart