SY3H3249FB81 Synthonix Corporation

1-Oxa-5-azaspiro[2.4]heptane-5-carboxylic acid tert-butyl ester

  • MDL number MFCD07779387
  • Cas Number : 301226-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6275.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/6275.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart