SY3H3249FB86 Synthonix Corporation

5-Oxa-2-aza-spiro[3.5]nonane hemioxalate

  • MDL number MFCD27988003
  • Cas Number : 1523571-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7196.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7196.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart