SY3H3249FB87 Synthonix Corporation

2-Boc-7-oxo-8-oxa-2,6-diaza-spiro[4.5]decane

  • MDL number MFCD22415198
  • Cas Number : 1363382-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7209.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7209.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart