SY3H3249FB88 Synthonix Corporation

8-Boc-2-oxo-3-oxa-1,8-diaza-spiro[5.5]undecane

  • MDL number MFCD22415208
  • Cas Number : 1363381-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7214.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7214.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart