SY3H3D6755AE Synthonix Corporation

2-(2-Aminothiazol-4-yl)acetic acid hydrochloride

  • MDL number MFCD06658943
  • Cas Number : 66659-20-9
  • Molecular Weight : 194.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39195.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39195.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart