SY3H3D6755BF Synthonix Corporation

(S)-2-Amino-2-(2-chlorophenyl)acetic acid

  • MDL number MFCD03426346
  • Cas Number : 141315-50-6
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39505.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39505.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart