SY3H3D6755C6 Synthonix Corporation

5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile

  • MDL number MFCD00227537
  • Cas Number : 120068-79-3
  • Molecular Weight : 321.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Very pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39666.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39666.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart